missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetyl-Histone H3 (Lys27) Rabbit anti-Human, Vertebrates, Yeast (S. cerevisiae ), Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer: Emd Millipore Corporation 07360S
Description
Anti-acetyl-Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody for detection of Histone H3 acetylated on lysine 27. Also known as Anti-H3K27ac this antibody is published in peer reviewed journals and is specificity verified by dot blot (DB) and validated in Chimmunoprecipitation, ChIP-seq, western blot, DB, Mplex.Specifications
acetyl-Histone H3 (Lys27) | |
Unconjugated | |
Serum | |
Rabbit | |
IgG | |
10μL | |
Primary | |
NM_003493 |
Chromatin Immunoprecipitation,Dot Blot,Immunoassay,Multiplexing,Western Blot | |
Immunoaffinity Purified immunoglobulin in 0.2M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide, and 5mg/ml BSA before the addition of glycerol to 30% | |
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886 | |
in which ACK corresponds to acetyl lysine 27 of yeast histone H3 | |
Unpurified | |
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles. | |
Polyclonal | |
Human,S. cerevisiae,Vertebrate,Yeast |
For Research Use Only