missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetyl-Histone H3 (Lys23) Rabbit anti-Human, Vertebrates, Yeast (S. cerevisiae), Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

Manufacturer:  Emd Millipore Corporation 07355

 View more versions of this product

Catalog No. 07-355-MI



Description

Description

Use Anti-acetyl-Histone H3 (Lys23) Antibody (Rabbit Polyclonal Antibody) validated in Chimmunoprecipitation, western blotting to detect acetyl-Histone H3 (Lys23) also known as H3K23Ac, Histone H3 (acetyl K23).
Specifications

Specifications

acetyl-Histone H3 (Lys23)
Unconjugated
Serum
Rabbit
Antiserum
−20°C for 24 months
Polyclonal
Human,S. cerevisiae,Vertebrate,Yeast
Chromatin Immunoprecipitation,Western Blot
Rabbit antiserum containing 0.05% sodium azide and 30% glycerol
HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t
Ovalbumin-conjugated
100μL
Primary
NM_003493
SDS
Documents
Product Certifications

For Research Use Only