missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetyl-Histone H3 (Lys18) Rabbit anti-Human, Vertebrates, Yeast (S. cerevisiae ), Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer: Emd Millipore Corporation 07354
Description
Use Anti-acetyl-Histone H3 (Lys18) Antibody (Rabbit Polyclonal Antibody) validated in Chimmunoprecipitation, western blotting to detect acetyl-Histone H3 (Lys18) also known as H3K18Ac, Histone H3 (acetyl K18).Specifications
acetyl-Histone H3 (Lys18) | |
Unconjugated | |
Serum | |
Rabbit | |
Unpurified | |
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles. | |
Polyclonal | |
Human,S. cerevisiae,Vertebrate,Yeast |
Chromatin Immunoprecipitation,Western Blot | |
100μLof rabbit serum containing 0.05% sodium azide and 30% glycerol. | |
HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t | |
KLH-conjugated | |
100μL | |
Primary | |
NM_003493 |
For Research Use Only