missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetyl-Histone H3 (Lys14) Rabbit anti-Human, Yeast (S. cerevisiae ), Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07353
Description
Anti-acetyl-Histone H3 (Lys14) Antibody is a rabbit polyclonal antibody for detection of acetyl-Histone H3 (Lys14) also known as H3K14Ac, Histone H3 (acetyl K14) and has been validated in Chimmunoprecipitation, western blot, ChIP-seq, DB, Mplex.Specifications
acetyl-Histone H3 (Lys14) | |
Polyclonal | |
Antiserum containing 0.05% sodium azide and 30% glycerol. | |
Rabbit | |
Unpurified | |
Primary | |
Human, S. cerevisiae, Yeast | |
Serum |
Multiplex, ChIP sequencing (ChIP-seq), Dot Blot, Immunoassay, Western Blot | |
Unconjugated | |
HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t | |
Ovalbumin-conjugated, synthetic peptide (KSTGGAcKAPRK-C) corresponding to amino acids 9-18 of yeast histone H3 acetylated on lysine 14, with a C-terminal cysteine added for conjugation purposes. | |
100 μL | |
NM_003493 | |
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles | |
IgG |
For Research Use Only