Please login to your online account to display your discounted pricing

anti-ACBD5, Polyclonal, Novus Biologicals™

$367.12 - $461.00

Specifications

Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:AQEEVKGAEQSDNDKKMMKKSADHKNLEVIVTNGYDKDGFVQDIQNDIHASSSLNGRSTEEVKPIDENLGQTGKSAVCIHQDINDDHVEDVTGIQHLTSDSDSEVYCDSMEQFGQEESLDSFTSNNGPFQYYLGGHSSQPM
Antigen ACBD5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP159820

 
novus biologicals canada
NBP159820
Unlabeled 0.05mg Each for $367.12

NBP185145

 
novus biologicals canada
NBP185145
Unlabeled 0.1mL Each for $461.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:AQEEVKGAEQSDNDKKMMKKSADHKNLEVIVTNGYDKDGFVQDIQNDIHASSSLNGRSTEEVKPIDENLGQTGKSAVCIHQDINDDHVEDVTGIQHLTSDSDSEVYCDSMEQFGQEESLDSFTSNNGPFQYYLGGHSSQPM
Antigen ACBD5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Gene Accession No. Q5T8D3
Gene ID (Entrez) 91452
Formulation PBS, pH 7.2, containing 40% glycerol