Please login to your online account to display your discounted pricing

anti-ACAT, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A fusion protein construct that contains the N-terminal region of human ACAT1. [UniProt No. P35610]
Antigen ACAT
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB400141

 
novus biologicals canada
NB400141
Unlabeled 0.1mL Each for $309.05

NB400141SS

 
novus biologicals canada
NB400141SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A fusion protein construct that contains the N-terminal region of human ACAT1. [UniProt No. P35610]
Antigen ACAT
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This product is specific for protein corresponding to ACAT1 - Sterol O-acyltransferase 1 (SOAT1) Uniprot# P35610, Gene ID# 6646. [Important Note:- ACAT1 is a symbol for both acyl-Coenzyme A: cholesterol acyltransferase 1 (SOAT1/ACAT1) and for acetyl-CoA acetyltransferase 1 (ACAT1/ THIL, Gene ID 38, Uniprot P24752) which often cause confusion].
Gene Accession No. P35610
Gene ID (Entrez) 6646