Please login to your online account to display your discounted pricing

anti-Abhd5, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region within residues 200-300 of the human Abhd5 protein. [Swiss-Prot No. Q8WTS1]
Antigen Abhd5
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11041576

 
novus biologicals canada
NB11041576
Unlabeled 0.1mL Each for $309.05

NB11041576T

 
novus biologicals canada
NB11041576SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region within residues 200-300 of the human Abhd5 protein. [Swiss-Prot No. Q8WTS1]
Antigen Abhd5
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Test Specificity Human and rat. Mouse reactivity reported in scientific literature (PMID:23878361).
Gene Accession No. Q8WTS1
Gene ID (Entrez) 51099
Formulation PBS