Please login to your online account to display your discounted pricing

anti-Abeta 42, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300226SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300226SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 351
Cross Reactivity Human
Cross Reactivity Mouse
Test Specificity Abeta 42. NB 300-226 reacts with human and mouse forms of the protein. There is minimal cross-reactivity with Abeta 40 (see Notes).
Host Species Rabbit
Conjugate Unlabeled
Antigen Abeta 42
Format Antiserum
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mL
Immunogen Whole Abeta 42 peptide.
Primary or Secondary Primary