Please login to your online account to display your discounted pricing

anti-Abeta 42, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300226

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300226

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 351
Cross Reactivity Human
Cross Reactivity Mouse
Primary or Secondary Primary
Test Specificity Abeta 42. NB 300-226 reacts with human and mouse forms of the protein. There is minimal cross-reactivity with Abeta 40 (see Notes).
Conjugate Unlabeled
Format Antiserum
Quantity 0.1mL
Applications Immunoprecipitation
Applications Western Blotting
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Antigen Abeta 42
Immunogen Whole Abeta 42 peptide.