Please login to your online account to display your discounted pricing

anti-Abeta 42, Polyclonal, Novus Biologicals™

$160.00 - $354.00

Specifications

Host Species Rabbit
Immunogen Whole Abeta 42 peptide.
Antigen Abeta 42
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB300226

 
novus biologicals canada
NB300226
Unlabeled 0.1mL Each for $354.00

NB300226SS

 
novus biologicals canada
NB300226SS
Unlabeled 0.025mL Each for $160.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Whole Abeta 42 peptide.
Antigen Abeta 42
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Abeta 42. NB 300-226 reacts with human and mouse forms of the protein. There is minimal cross-reactivity with Abeta 40 (see Notes).
Gene ID (Entrez) 351