Please login to your online account to display your discounted pricing

anti-Abeta 40, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acid residues GLMVGGVV of human Abeta [UniProt No. P05067]
Antigen Abeta 40
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB300225

 
novus biologicals canada
NB300225
Unlabeled 0.1mL Each for $329.00

NB300225SS

 
novus biologicals canada
NB300225SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acid residues GLMVGGVV of human Abeta [UniProt No. P05067]
Antigen Abeta 40
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Specific for Abeta 40; there is no cross-reactivity with Abeta 42.
Gene Accession No. P05067
Gene ID (Entrez) 351