Please login to your online account to display your discounted pricing

anti-Abeta 40, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300225SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300225SS

  • / Each
 

Description & Specifications

Specifications

Antigen Abeta 40
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Antisera
Gene Accession No. P05067
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acid residues GLMVGGVV of human Abeta [UniProt No. P05067]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 351
Test Specificity Specific for Abeta 40; there is no cross-reactivity with Abeta 42.
Target Species Human
Target Species Murine