Please login to your online account to display your discounted pricing

anti-ABCG8, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide from the N-terminal region of human ABCG8 protein. [UniProt No. Q9H221]
Antigen ABCG8
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB400110SS

 
novus biologicals canada
NB400110SS
Unlabeled 0.025mL Each for $139.00

NB400110

 
novus biologicals canada
NB400110
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide from the N-terminal region of human ABCG8 protein. [UniProt No. Q9H221]
Antigen ABCG8
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Human. Does not appear to react with mouse.
Gene Accession No. Q9H221
Gene ID (Entrez) 64241
Formulation PBS
Concentration 1mg/mL