Please login to your online account to display your discounted pricing

ABCE1 Polyclonal Antibody, Invitrogen™


ABCE1 Antibody, PA1-16797, from Invitrogen™. Species Reactivity: Human; Applications: Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot.

Manufacturer: pierce biotechnology inc  PA116797

Catalog No. APPA6797

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Formulation 100μL purified antibody (0.2mg/mL) in PBS and 0.05% sodium azide
Gene ID (Entrez) 6059
Isotype IgG
Gene Symbols ABCE1
Species Reactivity Human
Gene Alias ABC 38, ABC38, ABCE 1, ATP Binding Cassette Sub Family E Member 1, OABP, Ribonuclease 4 Inhibitor, RLI, RNase L Inhibitor, RNS4I, RNase 4 inhibitor
Gene Accession No. P61221
Storage Requirements Avoid Freeze/Thaw Cycles
Immunogen Fusion protein containing residues 1-119 of human ABCE1.
Cross Reactivity Human
Quantity 100μL
Host Species Rabbit
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Format Purified
Antigen ABCE1

ABCE1 Antibody, PA1-16797, from Invitrogen. Species Reactivity: Human; Applications: Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot.

ABCE1 is a member of the ABC Transporter superfamily, although it is not a membrane transporter. It is cytosolically expressed and inhibits the RNaseL protein. Recently, it has been found to be essential for the assembly of immature immunodeficiency virus capsids.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Fusion protein containing residues 1-119 of human ABCE1.
Formulation: 100μL purified antibody (0.2mg/mL) in PBS and 0.05% sodium azide
Synonyms: ABC 38, ABC38, ABCE 1, ATP Binding Cassette Sub Family E Member 1, OABP, Ribonuclease 4 Inhibitor, RLI, RNase L Inhibitor, RNS4I, RNase 4 inhibitor
Uniprot Accession: P61221
Entrez GeneID: 6059
Gene Symbol: ABCE1
Suggested positive control: antigen standard for ABCE1 (transient overexpression lysate), 293 whole cell lysate, Hela whole cell lysate, HeLa and 293 cell lysates.