Please login to your online account to display your discounted pricing

anti-53BP1, Polyclonal, Novus Biologicals™

$144.63 - $384.00

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residues 350 and 400 of human 53BP1 [NP_005648.1].
Antigen 53BP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100304SS

 
novus biologicals canada
NB100304SS
Unlabeled 0.025mL Each for $144.63

NB100304

 
novus biologicals canada
NB100304
Unlabeled 0.1mL Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residues 350 and 400 of human 53BP1 [NP_005648.1].
Antigen 53BP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Human, mouse, rat and goat. Human has been tested in WB, IHC-P and ICC/IF, mouse and goat have only been tested in ICC/IF. Primate reactivity reported in scientific literature (PMID: 18389475).Fish reactivity reported in scientific literature (PMID: 25516420).
Gene Accession No. Q12888
Gene ID (Entrez) 7158
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL