Please login to your online account to display your discounted pricing

anti-53BP1, Polyclonal, Novus Biologicals™

$164.00 - $384.00

Specifications

Host Species Rabbit
Immunogen 12 and 19 [UniProt No. Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305.
Antigen 53BP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100904SS

 
novus biologicals canada
NB100904SS
Unlabeled 0.025mL Each for $164.00

NB100904

 
novus biologicals canada
NB100904
Unlabeled 0.1mL Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen 12 and 19 [UniProt No. Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305.
Antigen 53BP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Human and mouse. Human has been tested in WB, IHC and ICC/IF, mouse has only been tested in ICC/IF.
Gene Accession No. Q12888
Gene ID (Entrez) 7158
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL