Please login to your online account to display your discounted pricing

anti-53BP1, HRP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100904H

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100904H

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Gene ID (Entrez) 7158
Test Specificity Human and mouse. Human has been tested in WB, IHC and ICC/IF, mouse has only been tested in ICC/IF.
Gene Accession No. Q12888
Conjugate HRP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Antigen 53BP1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 0.1mL
Immunogen 12 and 19 [UniProt No. Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305.
Primary or Secondary Primary
Host Species Rabbit
Regulatory Status RUO