anti-53BP1, FITC, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals Canada NB100304F
 View more versions of this product
Catalog No. NB100304F
Disclaimers:
For Research Use Only
Specifications Description & Specifications
Specifications
53BP1
1mg/mL
Fish,Goat,Human,Mouse,Primate,Rat
50mM sodium borate
Rabbit
0.1mL
Primary
7158
Fish,Goat,Human,Murine,Primate,Rat
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Western Blot
FITC
Affinity Purified
Q12888
The epitope recognized by this antibody maps to a region between residues 350 and 400 of human 53BP1 [NP_005648.1].
RUO
Polyclonal
Human, mouse, rat and goat. Human has been tested in WB, IHC-P and ICC/IF, mouse and goat have only been tested in ICC/IF. Primate reactivity reported in scientific literature (PMID: 18389475).Fish reactivity reported in scientific literature (PMID: 25516420).