Please login to your online account to display your discounted pricing

anti-53BP1, FITC, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100904F

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100904F

  • / Each
 

Description & Specifications

Specifications

Formulation 50mM sodium borate
Gene Accession No. Q12888
Test Specificity Human and mouse. Human has been tested in WB, IHC and ICC/IF, mouse has only been tested in ICC/IF.
Monoclonal or Polyclonal Polyclonal
Conjugate FITC
Gene ID (Entrez) 7158
Quantity 0.1mL
Format Affinity Purified
Antigen 53BP1
Immunogen 12 and 19 [UniProt No. Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305.
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Host Species Rabbit
Regulatory Status RUO
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse