Please login to your online account to display your discounted pricing

anti-53BP1, DyLight 650, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100904C

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100904C

  • / Each
 

Description & Specifications

Specifications

Immunogen 12 and 19 [UniProt No. Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305.
Gene Accession No. Q12888
Test Specificity Human and mouse. Human has been tested in WB, IHC and ICC/IF, mouse has only been tested in ICC/IF.
Cross Reactivity Human
Cross Reactivity Mouse
Formulation 50mM sodium borate
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7158
Host Species Rabbit
Antigen 53BP1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Regulatory Status RUO
Primary or Secondary Primary
Conjugate DyLight 650
Format Affinity Purified
Quantity 0.1mL