Please login to your online account to display your discounted pricing

anti-53BP1, DyLight 550, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100904R

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100904R

  • / Each
 

Description & Specifications

Specifications

Formulation 50mM sodium borate
Gene Accession No. Q12888
Primary or Secondary Primary
Conjugate DyLight 550
Monoclonal or Polyclonal Polyclonal
Test Specificity Human and mouse. Human has been tested in WB, IHC and ICC/IF, mouse has only been tested in ICC/IF.
Format Affinity Purified
Gene ID (Entrez) 7158
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen 12 and 19 [UniProt No. Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305.
Antigen 53BP1
Regulatory Status RUO
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rabbit