Please login to your online account to display your discounted pricing

anti-53BP1, DyLight 405, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100304V

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100304V

  • / Each
 

Description & Specifications

Specifications

Formulation 50mM sodium borate
Gene ID (Entrez) 7158
Test Specificity Human, mouse, rat and goat. Human has been tested in WB, IHC-P and ICC/IF, mouse and goat have only been tested in ICC/IF. Primate reactivity reported in scientific literature (PMID: 18389475).Fish reactivity reported in scientific literature (PMID: 25516420).
Cross Reactivity Fish
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Primary or Secondary Primary
Concentration 1mg/mL
Gene Accession No. Q12888
Immunogen The epitope recognized by this antibody maps to a region between residues 350 and 400 of human 53BP1 [NP_005648.1].
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Quantity 0.1mL
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Antigen 53BP1
Format Affinity Purified
Conjugate DyLight 405
Host Species Rabbit