anti-53BP1, DyLight 405, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals Canada NB100904V
 View more versions of this product
Catalog No. NB100904V
Disclaimers:
For Research Use Only
Specifications Description & Specifications
Specifications
53BP1
DyLight 405
Affinity Purified
Q12888
12 and 19 [UniProt No. Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305.
RUO
Polyclonal
Human and mouse. Human has been tested in WB, IHC and ICC/IF, mouse has only been tested in ICC/IF.
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
Human,Mouse
50mM sodium borate
Rabbit
0.1mL
Primary
7158
Human,Murine