Please login to your online account to display your discounted pricing

anti-53BP1, Biotin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100304B

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100304B

  • / Each
 

Description & Specifications

Specifications

Antigen 53BP1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Conjugate Biotin
Cross Reactivity Fish
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS
Gene Accession No. Q12888
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residues 350 and 400 of human 53BP1 [NP_005648.1].
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7158
Test Specificity Human, mouse, rat and goat. Human has been tested in WB, IHC-P and ICC/IF, mouse and goat have only been tested in ICC/IF. Primate reactivity reported in scientific literature (PMID: 18389475).Fish reactivity reported in scientific literature (PMID: 25516420).
Target Species Fish
Target Species Goat
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat