Please login to your online account to display your discounted pricing

anti-53BP1, Alexa Fluor 647, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100904AF647

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100904Y

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q12888
Primary or Secondary Primary
Test Specificity Human and mouse. Human has been tested in WB, IHC and ICC/IF, mouse has only been tested in ICC/IF.
Formulation 50mM Sodium Borate
Gene ID (Entrez) 7158
Conjugate Alexa Fluor 647
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Immunogen 12 and 19 [UniProt No. Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305.
Antigen 53BP1
Format Affinity Purified
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 100 Tests
Regulatory Status RUO