Please login to your online account to display your discounted pricing

anti-53BP1, Alexa Fluor 405, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100304AF405

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100304W

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7158
Conjugate Alexa Fluor 405
Regulatory Status RUO
Format Affinity Purified
Cross Reactivity Fish
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Formulation 50mM Sodium Borate
Gene Accession No. Q12888
Test Specificity Human, mouse, rat and goat. Human has been tested in WB, IHC-P and ICC/IF, mouse and goat have only been tested in ICC/IF. Primate reactivity reported in scientific literature (PMID: 18389475).Fish reactivity reported in scientific literature (PMID: 25516420).
Quantity 100 Tests
Antigen 53BP1
Monoclonal or Polyclonal Polyclonal
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residues 350 and 400 of human 53BP1 [NP_005648.1].