Please login to your online account to display your discounted pricing

anti-53BP1, Alexa Fluor 405, Polyclonal, Novus Biologicals™

$428.00 - $428.00

Specifications

Host Species Rabbit
Immunogen 12 and 19 [UniProt No. Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305.
Antigen 53BP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100304W

 
novus biologicals canada
NB100304AF405
Alexa Fluor 405 100 Tests Each for $428.00

NB100904W

 
novus biologicals canada
NB100904AF405
Alexa Fluor 405 100 Tests Each for $428.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen 12 and 19 [UniProt No. Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305.
Antigen 53BP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human and mouse. Human has been tested in WB, IHC and ICC/IF, mouse has only been tested in ICC/IF.
Gene Accession No. Q12888
Gene ID (Entrez) 7158
Formulation 50mM Sodium Borate