Please login to your online account to display your discounted pricing

anti-5-Lipoxygenase, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region within residues 600-674 of human 5-Lipoxygenase. [Swiss-Prot No. P09917]
Antigen 5-Lipoxygenase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11058749T

 
novus biologicals canada
NB11058749SS
Unlabeled 0.025mL Each for $109.00

NB11058749

 
novus biologicals canada
NB11058749
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region within residues 600-674 of human 5-Lipoxygenase. [Swiss-Prot No. P09917]
Antigen 5-Lipoxygenase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blotting
Gene Accession No. P09917
Gene ID (Entrez) 240
Formulation PBS, 30% glycerol
Concentration 1mg/mL