Please login to your online account to display your discounted pricing

anti-5-Lipoxygenase, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a region within residues 100-200 of human 5-Lipoxygenase. [Swiss-Prot No. P09917]
Antigen 5-Lipoxygenase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11058748T

 
novus biologicals canada
NB11058748SS
Unlabeled 0.025mL Each for $139.00

NB11058748

 
novus biologicals canada
NB11058748
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a region within residues 100-200 of human 5-Lipoxygenase. [Swiss-Prot No. P09917]
Antigen 5-Lipoxygenase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Gene Accession No. P09917
Gene ID (Entrez) 240
Formulation PBS, 30% glycerol
Concentration 1mg/mL