Please login to your online account to display your discounted pricing

anti-5-Lipoxygenase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11058749SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11058749T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P09917
Formulation PBS, 30% glycerol
Concentration 1mg/mL
Immunogen A synthetic peptide made to a C-terminal region within residues 600-674 of human 5-Lipoxygenase. [Swiss-Prot No. P09917]
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 240
Conjugate Unlabeled
Quantity 0.025mL
Antigen 5-Lipoxygenase
Format Affinity Purified
Host Species Rabbit
Regulatory Status RUO
Applications Simple Western
Applications Western Blotting
Primary or Secondary Primary