Please login to your online account to display your discounted pricing

anti-5-Lipoxygenase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11058749

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11058749

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 240
Quantity 0.1mL
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Conjugate Unlabeled
Formulation PBS, 30% glycerol
Concentration 1mg/mL
Gene Accession No. P09917
Immunogen A synthetic peptide made to a C-terminal region within residues 600-674 of human 5-Lipoxygenase. [Swiss-Prot No. P09917]
Applications Simple Western
Applications Western Blotting
Host Species Rabbit
Regulatory Status RUO
Antigen 5-Lipoxygenase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary