Please login to your online account to display your discounted pricing

anti-5-HT7, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Immunogen This antibody was developed by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 13-28 of the rat 5-HT7R (AAA42134.1).
Antigen 5-HT7
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056352T

 
novus biologicals canada
NB10056352SS
Unlabeled 0.025mg Each for $139.00

NB10056352

 
novus biologicals canada
NB10056352
Unlabeled 0.1mg Each for $362.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 13-28 of the rat 5-HT7R (AAA42134.1).
Antigen 5-HT7
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity This sequence is identical for 5-HT7R splice variants in the rat (5-HT7Ra/b/c), human (5-HT7Ra/b/d) and human 5-HT7, the mouse 5-HT7R. It is 93% conserved with dog 5-HT7Ra/b, and 81% conserved with pig 5-HT7R. The 5HT7 antibody recognizes all described 5HT7 receptor splice variants. Reacts with Dog, Human, Mouse and Rat. Predicted to react with Porcine.
Gene ID (Entrez) 3363
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL