Please login to your online account to display your discounted pricing

anti-5-HT3A, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Immunogen This antibody was developed by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 27-42 (RATQAHSTTQPALLRL) and 427-442 (LSSIRHSLEKRDEMRE) of rat 5-HT3 Receptor
Antigen 5-HT3A
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056351

 
novus biologicals canada
NB10056351
Unlabeled 0.1mg Each for $362.00

NB10056351T

 
novus biologicals canada
NB10056351SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 27-42 (RATQAHSTTQPALLRL) and 427-442 (LSSIRHSLEKRDEMRE) of rat 5-HT3 Receptor
Antigen 5-HT3A
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity In human brain, a 47-50 kDa band is obeserved.
Gene ID (Entrez) 3359
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL