Please login to your online account to display your discounted pricing

anti-5-HT1D, Polyclonal, Novus Biologicals™

$139.00 - $274.59

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 1-18 and 251-267 of rat 5-HT1DR (Genbank accession no. NP_036984).
Antigen 5-HT1D
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056349

 
novus biologicals canada
NB10056349
Unlabeled 0.1mg Each for $274.59

NB10056349T

 
novus biologicals canada
NB10056349SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 1-18 and 251-267 of rat 5-HT1DR (Genbank accession no. NP_036984).
Antigen 5-HT1D
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blotting
Gene ID (Entrez) 3352
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL