Please login to your online account to display your discounted pricing

anti-5-HT1D, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056349

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056349

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Gene ID (Entrez) 3352
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Monoclonal or Polyclonal Polyclonal
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL
Conjugate Unlabeled
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 1-18 and 251-267 of rat 5-HT1DR (Genbank accession no. NP_036984).
Applications Simple Western
Applications Western Blotting
Regulatory Status RUO
Quantity 0.1mg
Antigen 5-HT1D
Primary or Secondary Primary
Format Purified