Please login to your online account to display your discounted pricing

anti-4933415F23Rik, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227164

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227164

  • / Each
 

Description & Specifications

Specifications

Antigen 4933415F23Rik
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Gene Accession No. Q14BX6
Host Species Rabbit
Immunogen A portion of amino acids 20-93 of mouse protein was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 66755
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse and 64% homologous in rat.
Target Species Human