Please login to your online account to display your discounted pricing

anti-4933415F23Rik, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 20-93 of mouse protein was used as the immunogen.
Antigen 4933415F23Rik
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227164

 
novus biologicals canada
NBP227164
Unlabeled 0.1mg Each for $255.80

NBP227164SS

 
novus biologicals canada
NBP227164SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 20-93 of mouse protein was used as the immunogen.
Antigen 4933415F23Rik
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse and 64% homologous in rat.
Gene Accession No. Q14BX6
Gene ID (Entrez) 66755
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL