Please login to your online account to display your discounted pricing

anti-4933415F23Rik, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227164SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227164SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene ID (Entrez) 66755
Concentration 0.5mg/mL
Gene Accession No. Q14BX6
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse and 64% homologous in rat.
Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Monoclonal or Polyclonal Polyclonal
Immunogen A portion of amino acids 20-93 of mouse protein was used as the immunogen.
Cross Reactivity Human
Format Purified
Quantity 0.025mg
Host Species Rabbit
Antigen 4933415F23Rik
Applications Western Blotting
Regulatory Status RUO