missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-beta-2 Glycoprotein-1, Polyclonal, Invitrogen™

beta-2 Glycoprotein-1 Polyclonal antibody PA1-74015 from Thermo Scientific™ specifically detects beta-2 Glycoprotein-1 in Canine, Human, Porcine and Rat samples. It is validated for ELISA.

Manufacturer:  Pierce Biotechnology Inc PA174015

Catalog No. PIPA174015


 

Specifications

Description

beta-2 Glycoprotein-1 Polyclonal antibody PA1-74015 from Thermo Scientific™ specifically detects beta-2 Glycoprotein-1 in Canine, Human, Porcine and Rat samples. It is validated for ELISA.

Host Species: Goat
Cross Reactivity: Canine, Human, Porcine, Rat
Isotype: IgG
Immunogen: Apolipoprotein-H from human plasma.
Formulation: 5mg Purified Antibody in 0.01M HEPES, with 0.15M NaCl, 50% glycerol and no preservative; pH7.4
Uniprot Accession: P02749, P33703
Entrez GeneID: 100522100, 287774, 350, 403945
Gene Symbol: APOH
PA1-74015 detects beta-2 Glycoprotein-1 from human, rat, canine, and porcine samples.PA1-74015 has been successfully used in ELISA and immunoprecipitation applications.The PA1-74015 immunogen is Apolipoprotein-H from human plasma.
Description & Specifications

Specifications

beta-2 Glycoprotein-1
Unlabeled
P02749, P33703
APOH
Apolipoprotein-H from human plasma.
5mg
Primary
100522100, 287774, 350, 403945
ELISA
5 mg purified Antibody in 0.01M HEPES, with 0.15M NaCl, 50% glycerol and no preservative; 7.4
Purified
Goat
IgG
Avoid Freeze/Thaw Cycles
Polyclonal
Canine,Human,Porcine,Rat
Promotions

Promotions

  • Purchase any three Invitrogen™ Antibodies and receive a fourth antibody at no additional cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.

    06/30/18