Please login to your online account to display your discounted pricing

anti-15-PGDH/HPGD, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Immunogen Purified type I human placental 15-PGDH protein [UniProt No. P15428]
Antigen 15-PGDH/HPGD
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB200179SS

 
novus biologicals canada
NB200179SS
Unlabeled 0.025mL Each for $139.00

NB200179

 
novus biologicals canada
NB200179
Unlabeled 0.1mL Each for $362.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Purified type I human placental 15-PGDH protein [UniProt No. P15428]
Antigen 15-PGDH/HPGD
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Gene Accession No. P15428
Gene ID (Entrez) 3248
Formulation Tris-glycine, 150mM NaCl
Concentration 2mg/mL