Please login to your online account to display your discounted pricing

anti-14-3-3 zeta, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 200-245 of human 14-3-3 zeta was used as the immunogen for the antibody.
Antigen 14-3-3 zeta
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224593

 
novus biologicals canada
NBP224593
Unlabeled 0.1mg Each for $255.80

NBP224593SS

 
novus biologicals canada
NBP224593SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 200-245 of human 14-3-3 zeta was used as the immunogen for the antibody.
Antigen 14-3-3 zeta
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human and 94% homologous in mouse, rat and bovine.
Gene ID (Entrez) 7534
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL