Please login to your online account to display your discounted pricing

anti-14-3-3 zeta, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224595SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224595SS

  • / Each
 

Description & Specifications

Specifications

Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and dog and 93% homologous in mouse, rat and monkey.
Concentration 0.5mg/mL
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Porcine
Cross Reactivity Primate
Formulation 100μg in 200μL PBS containing 0.05% BSA
Regulatory Status RUO
Gene ID (Entrez) 7534
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A portion of amino acids 50-100 of human 14-3-3 zeta was used as the immunogen.
Antigen 14-3-3 zeta
Quantity 0.025mg
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Format Purified
Primary or Secondary Primary
Conjugate Unlabeled