Please login to your online account to display your discounted pricing

anti-14-3-3 zeta, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224595

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224595

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7534
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and dog and 93% homologous in mouse, rat and monkey.
Host Species Rabbit
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Porcine
Cross Reactivity Primate
Formulation 100μg in 200μL PBS containing 0.05% BSA
Conjugate Unlabeled
Concentration 0.5mg/mL
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A portion of amino acids 50-100 of human 14-3-3 zeta was used as the immunogen.
Primary or Secondary Primary
Antigen 14-3-3 zeta
Quantity 0.1mg
Format Purified