Please login to your online account to display your discounted pricing

anti-14-3-3, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 40-90 from human 14-3-3 was used as the immunogen for this antibody.
Antigen 14-3-3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227370SS

 
novus biologicals canada
NBP227370SS
Unlabeled 0.025mg Each for $109.00

NBP227370

 
novus biologicals canada
NBP227370
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 40-90 from human 14-3-3 was used as the immunogen for this antibody.
Antigen 14-3-3
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, crab-eating macaque, and rhesus monkey.
Gene Accession No. P27348
Gene ID (Entrez) 10971
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL