anti-14-3-3γ, Clone: CG31-2B6, MilliporeSigma™
Manufacturer:  Emd Millipore Corporation 05639
Catalog No. 05-639-MI
Description
Description
Specifications
Specifications
14-3-3γ
CG31-2B6
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Mouse
IgG1
100μg
Primary
Human,Murine,Rat
Western Blot
Unconjugated
NM_012479
YWHAG, KCIP-1, 14-3-3GAMMA
GST fusion protein corresponding to full-length human 14-3-3 gamma
Protein A chromatography
−20°C for 24 months.
Monoclonal