Please login to your online account to display your discounted pricing

Anthrone, For analysis ACS, ACROS Organics™


Quantity 500g
Packaging Plastic Bottle

Manufacturer: thermo fisher scientific  AC104965000

 View more versions of this product

Catalog No. AC104965000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Crystals
Formula Weight 194.23
Quantity 500g
Packaging Plastic Bottle
Boiling Point 721.0°C
Color Yellow
Solubility Solubility in water: insoluble. Other solubilities: soluble in hot naoh solution, conc. sulfuric acid, soluble in alcohol, benzene, aceton, soluble in most organic solvents without, fluorescence
Melting Point 152° to 157°C
Synonym 9(10H)-Anthracenone
Absorbance 0.75 max. (425nm, 1cm cell) Passes Test, 0.045 max. (620nm, 1cm cell) Passes Test
Chemical Name or Material Anthrone
Infrared Spectrum Authentic
Molecular Formula C14H10O
Sensitivity (to carbohydrates) Passes Test
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00001187
CAS 90-44-8

Specifications

Chemical Name or Material Anthrone
CAS 90-44-8
Purity Grade ACS Reagent Grade
Molecular Formula C14H10O
Formula Weight 194.23
MDL Number MFCD00001187
Synonym 9(10H)-Anthracenone
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents