missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Anthrone, 95%, pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 401365000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Anthrone | |
721.0°C | |
Glass bottle | |
MFCD00001187 | |
07,473 | |
anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744 | |
RJGDLRCDCYRQOQ-UHFFFAOYSA-N | |
10H-anthracen-9-one | |
7018 | |
194.23 | |
Pure |
90-44-8 | |
94% min. (GC) | |
C14H10O | |
500g | |
15,682 | |
Solubility in water: insoluble. Other solubilities: soluble in hot naoh solution, conc. sulfuric acid, soluble in alcohol, benzene, aceton, soluble in most organic solvents without, fluorescence | |
O=C1C2=CC=CC=C2CC2=CC=CC=C12 | |
194.23 | |
CHEBI:33835 | |
95% |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 201-994-
RUO â Research Use Only