Please login to your online account to display your discounted pricing

Anthraquinone 98%, ACROS Organics™

$36.90 - $93.79

Chemical Identifiers

Chemical Name or Material Anthraquinone
CAS 84-65-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H8O2
Formula Weight 208.22
MDL Number MFCD00001188
Synonym 9, 10-Dioxoanthracene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC104930500

  SDS
fisher scientific acros
AC104930500
Plastic bottle 50g Each for $36.90

AC104930010

  SDS
fisher scientific acros
104930010
Plastic bottle 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC104932500

  SDS
fisher scientific acros
104932500
Plastic bottle 250g Each for $93.79
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 208.22
Quantity 50g
Color Yellow-green to khaki to tan
Melting Point 283° to 288°C
Beilstein 07, 781
Fieser 04,20
Merck Index 15, 679
MDL Number MFCD00001188
Assay Percent Range 98%
Health Hazard 2 GHS P Statement May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Wear eye protection/face protection.
Packaging Plastic bottle
Synonym 9, 10-Dioxoanthracene
Molecular Formula C14H8O2
Infrared Spectrum Authentic
Chemical Name or Material Anthraquinone
CAS 84-65-1
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Wear eye protection/face protection.

GHS P Statement May cause an allergic skin reaction.