missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Anthraquinone-2-carboxylic acid, 98%

$70.03 - $240.57

Chemical Identifiers

CAS 117-78-2
Molecular Formula C15H8O4
Molecular Weight (g/mol) 252.225
MDL Number MFCD00001231
InChI Key ASDLSKCKYGVMAI-UHFFFAOYSA-N
Synonym anthraquinone-2-carboxylic acid, 9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, 2-carboxyanthraquinone, 2-anthraquinonecarboxylic acid, 2-carboxyl-9,10-anthraquinone, anthraquinone-2-carboxylate, 9,10-dihydro-9,10-dioxo-2-anthracenecarboxylic acid, 9,10-anthraquinone 2-carboxylic acid, .beta.-anthraquinonecarboxylic acid, anthraquinone-.beta.-carboxylic acid
PubChem CID 67030
IUPAC Name 9,10-dioxoanthracene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6092903 View Documents Alfa Aesar™
H6092903
1g Each for $70.03
 
AAH6092906 View Documents Alfa Aesar™
H6092906
5g Each for $240.57
 
Description

Description

Applications
Anthraquinone 2-carboxylic acid (AQ) as a novel electron shuttling mediator and attached electron relay for HRP. Efficient method for the generation of hydrogen peroxide by aerobic photooxidation of 2-propanol using anthraquinone-2-carboxylic acid and molecular oxygen is investigated.

Solubility
Soluble in hot Acetic acid(almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

117-78-2
252.225
ASDLSKCKYGVMAI-UHFFFAOYSA-N
67030
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)C(=O)O
C15H8O4
MFCD00001231
anthraquinone-2-carboxylic acid, 9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, 2-carboxyanthraquinone, 2-anthraquinonecarboxylic acid, 2-carboxyl-9,10-anthraquinone, anthraquinone-2-carboxylate, 9,10-dihydro-9,10-dioxo-2-anthracenecarboxylic acid, 9,10-anthraquinone 2-carboxylic acid, .beta.-anthraquinonecarboxylic acid, anthraquinone-.beta.-carboxylic acid
9,10-dioxoanthracene-2-carboxylic acid

Specifications

287°C to 289°C
117-78-2
MFCD00001231
ASDLSKCKYGVMAI-UHFFFAOYSA-N
9,10-dioxoanthracene-2-carboxylic acid
67030
98%
1g
C15H8O4
anthraquinone-2-carboxylic acid, 9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, 2-carboxyanthraquinone, 2-anthraquinonecarboxylic acid, 2-carboxyl-9,10-anthraquinone, anthraquinone-2-carboxylate, 9,10-dihydro-9,10-dioxo-2-anthracenecarboxylic acid, 9,10-anthraquinone 2-carboxylic acid, .beta.-anthraquinonecarboxylic acid, anthraquinone-.beta.-carboxylic acid
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)C(=O)O
252.225
252.22
Anthraquinone-2-carboxylic acid