missing translation for 'onlineSavingsMsg'
Learn More

Anthracene, Practical, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor A129625KG

 View more versions of this product

Catalog No. 18608885


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

120-12-7
215° to 219°C
Poly Pail
Practical
1
342°C
2.5 kg
Crystalline Powder or Lumps

Chemical Identifiers

120-12-7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.